PRIH8BA0582B Princeton BioMolecular Research, Inc.

4-amino-N-ethyl-N-phenyl-1-propyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417846
  • Cas Number : 2101195-47-3
  • Molecular Weight : 272.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart