PRIH8BA0582F Princeton BioMolecular Research, Inc.

4-amino-1-cyclopentyl-N-cyclopropyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417850
  • Cas Number : 2101195-79-1
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart