PRIH8BA05838 Princeton BioMolecular Research, Inc.

4-amino-1-cyclopentyl-N-(tetrahydrofuran-2-ylmethyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417859
  • Cas Number : 2101198-99-4
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart