PRIH8BA05843 Princeton BioMolecular Research, Inc.

4-amino-1-cyclopentyl-N,N-diethyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417870
  • Cas Number : 2101198-51-8
  • Molecular Weight : 250.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart