PRIH8BA05846 Princeton BioMolecular Research, Inc.

1-cyclopentyl-3-[(2-methylpiperidin-1-yl)carbonyl]-1H-pyrazol-4-amine

  • MDL number MFCD31417873
  • Cas Number : 2101199-76-0
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart