PRIH8BA0585D Princeton BioMolecular Research, Inc.

4-amino-N-[2-(dimethylamino)ethyl]-1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417895
  • Cas Number : 2101198-72-3
  • Molecular Weight : 279.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart