PRIH8BA0585F Princeton BioMolecular Research, Inc.

4-amino-N-[3-(dimethylamino)propyl]-1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417897
  • Cas Number : 2101195-84-8
  • Molecular Weight : 293.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart