PRIH8BA05865 Princeton BioMolecular Research, Inc.

3-(pyrrolidin-1-ylcarbonyl)-1-(2,2,2-trifluoroethyl)-1H-pyrazol-4-amine

  • MDL number MFCD31417903
  • Cas Number : 2101198-83-6
  • Molecular Weight : 262.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart