PRIH8BA0586D Princeton BioMolecular Research, Inc.

3-[(2-ethylpiperidin-1-yl)carbonyl]-1-(2,2,2-trifluoroethyl)-1H-pyrazol-4-amine

  • MDL number MFCD31417911
  • Cas Number : 2101198-78-9
  • Molecular Weight : 304.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart