PRIH8BA0586F Princeton BioMolecular Research, Inc.

4-amino-N-ethyl-N-phenyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417913
  • Cas Number : 2101198-82-5
  • Molecular Weight : 312.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart