PRIH8BA05892 Princeton BioMolecular Research, Inc.

4-amino-N-cyclopropyl-1-(2-fluoroethyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417949
  • Cas Number : 2101198-44-9
  • Molecular Weight : 212.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart