PRIH8BA05898 Princeton BioMolecular Research, Inc.

4-amino-1-(2-fluoroethyl)-N-(2-methoxyethyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417955
  • Cas Number : 2101196-08-9
  • Molecular Weight : 230.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart