PRIH8BA058A1 Princeton BioMolecular Research, Inc.

4-amino-1-(2-fluoroethyl)-N-(2-furylmethyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417959
  • Cas Number : 2101199-67-9
  • Molecular Weight : 252.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart