PRIH8BA058AA Princeton BioMolecular Research, Inc.

4-amino-N,N-diethyl-1-(2-fluoroethyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417969
  • Cas Number : 2101199-68-0
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart