PRIH8BA058AC Princeton BioMolecular Research, Inc.

1-(2-fluoroethyl)-3-(piperidin-1-ylcarbonyl)-1H-pyrazol-4-amine

  • MDL number MFCD31417971
  • Cas Number : 2101197-11-7
  • Molecular Weight : 240.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart