PRIH8BA058B5 Princeton BioMolecular Research, Inc.

4-amino-N-isopropyl-1-methyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD21194608
  • Cas Number : 1502548-69-7
  • Molecular Weight : 182.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart