PRIH8BA058CD Princeton BioMolecular Research, Inc.

4-amino-N-[2-(dimethylamino)ethyl]-1-ethyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD21221765
  • Cas Number : 1495005-42-9
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart