PRIH8BA058D3 Princeton BioMolecular Research, Inc.

4-amino-N-[3-(diethylamino)propyl]-1-ethyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD20796606
  • Cas Number : 1482663-28-4
  • Molecular Weight : 267.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart