PRIH8BA058E0 Princeton BioMolecular Research, Inc.

4-amino-N-ethyl-1-isopropyl-N-methyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD31417989
  • Cas Number : 2101195-83-7
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart