PRIH8BA058FB Princeton BioMolecular Research, Inc.

4-amino-1-isopropyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418007
  • Cas Number : 1644041-11-1
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart