PRIH8BA05909 Princeton BioMolecular Research, Inc.

4-amino-N-(2-furylmethyl)-1-propyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418021
  • Cas Number : 2101196-72-7
  • Molecular Weight : 248.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart