PRIH8BA05917 Princeton BioMolecular Research, Inc.

4-amino-1-propyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD24645087
  • Cas Number : 1357087-12-7
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart