PRIH8BA05919 Princeton BioMolecular Research, Inc.

4-amino-1-cyclopentyl-N-ethyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418036
  • Cas Number : 2101198-81-4
  • Molecular Weight : 222.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart