PRIH8BA0591D Princeton BioMolecular Research, Inc.

4-amino-1-cyclopentyl-N-(2-hydroxyethyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418040
  • Cas Number : 2101198-93-8
  • Molecular Weight : 238.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart