PRIH8BA0591F Princeton BioMolecular Research, Inc.

4-amino-1-cyclopentyl-N-ethyl-N-methyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418042
  • Cas Number : 2101200-93-3
  • Molecular Weight : 236.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart