PRIH8BA05923 Princeton BioMolecular Research, Inc.

4-amino-N-cyclohexyl-1-cyclopentyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418046
  • Cas Number : 2101196-26-1
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart