PRIH8BA05924 Princeton BioMolecular Research, Inc.

4-amino-1-cyclopentyl-N-(tetrahydrofuran-2-ylmethyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418047
  • Cas Number : 2101196-19-2
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart