PRIH8BA0592C Princeton BioMolecular Research, Inc.

4-amino-1-cyclopentyl-N-[3-(diethylamino)propyl]-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418055
  • Cas Number : 2101196-34-1
  • Molecular Weight : 307.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart