PRIH8BA0592D Princeton BioMolecular Research, Inc.

4-amino-1-cyclopentyl-N-(3-morpholin-4-ylpropyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418056
  • Cas Number : 2101199-00-0
  • Molecular Weight : 321.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart