PRIH8BA05932 Princeton BioMolecular Research, Inc.

5-(azepan-1-ylcarbonyl)-1-cyclopentyl-1H-pyrazol-4-amine

  • MDL number MFCD31418061
  • Cas Number : 2101196-25-0
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart