PRIH8BA05942 Princeton BioMolecular Research, Inc.

4-amino-N-(tetrahydrofuran-2-ylmethyl)-1-(2,2,2-trifluoroethyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418075
  • Cas Number : 2101199-25-9
  • Molecular Weight : 292.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart