PRIH8BA05945 Princeton BioMolecular Research, Inc.

4-amino-N-(3-methoxypropyl)-1-(2,2,2-trifluoroethyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418078
  • Cas Number : 2101200-96-6
  • Molecular Weight : 280.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart