PRIH8BA0594D Princeton BioMolecular Research, Inc.

4-amino-N,N-diethyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418086
  • Cas Number : 2101200-98-8
  • Molecular Weight : 264.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart