PRIH8BA05957 Princeton BioMolecular Research, Inc.

4-amino-1-(2,2-difluoroethyl)-N-(2-hydroxyethyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418096
  • Cas Number : 2101196-47-6
  • Molecular Weight : 234.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart