PRIH8BA05962 Princeton BioMolecular Research, Inc.

4-amino-1-(2,2-difluoroethyl)-N-[3-(dimethylamino)propyl]-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418107
  • Cas Number : 2101199-89-5
  • Molecular Weight : 275.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart