PRIH8BA05968 Princeton BioMolecular Research, Inc.

1-(2,2-difluoroethyl)-5-(pyrrolidin-1-ylcarbonyl)-1H-pyrazol-4-amine

  • MDL number MFCD31418113
  • Cas Number : 2101199-45-3
  • Molecular Weight : 244.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart