PRIH8BA0596F Princeton BioMolecular Research, Inc.

4-amino-1-(2-fluoroethyl)-N-methyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418119
  • Cas Number : 2101197-19-5
  • Molecular Weight : 186.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart