PRIH8BA05975 Princeton BioMolecular Research, Inc.

4-amino-N-butyl-1-(2-fluoroethyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418125
  • Cas Number : 2101196-78-3
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart