PRIH8BA05979 Princeton BioMolecular Research, Inc.

4-amino-N-cyclopentyl-1-(2-fluoroethyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418129
  • Cas Number : 2101196-79-4
  • Molecular Weight : 240.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart