PRIH8BA0597A Princeton BioMolecular Research, Inc.

4-amino-N-cyclohexyl-1-(2-fluoroethyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD31418130
  • Cas Number : 2101196-84-1
  • Molecular Weight : 254.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart