PRIH8BA05994 Princeton BioMolecular Research, Inc.

(4-bromo-1-propyl-1H-pyrazol-3-yl)methanol

  • MDL number MFCD31418153
  • Cas Number : 2101199-64-6
  • Molecular Weight : 219.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart