PRIH8BA059B1 Princeton BioMolecular Research, Inc.

(4-bromo-1-sec-butyl-1H-pyrazol-5-yl)methanol

  • MDL number MFCD31418180
  • Cas Number : 2101197-25-3
  • Molecular Weight : 233.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart