PRIH8BA059B6 Princeton BioMolecular Research, Inc.

(4-iodo-1-isobutyl-1H-pyrazol-5-yl)methanol

  • MDL number MFCD31418185
  • Cas Number : 2101195-06-4
  • Molecular Weight : 280.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart