PRIH8BA059BB Princeton BioMolecular Research, Inc.

[1-(2,2-difluoroethyl)-4-iodo-1H-pyrazol-5-yl]methanol

  • MDL number MFCD31418190
  • Cas Number : 2101197-45-7
  • Molecular Weight : 288.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart