PRIH8BA059BF Princeton BioMolecular Research, Inc.

4-amino-1-methyl-N-(tetrahydrofuran-2-ylmethyl)-1H-pyrazole-3-carboxamide

  • Cas Number : 1352201-48-9
  • Molecular Weight : 183.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart