PRIH8BA05A0B Princeton BioMolecular Research, Inc.

5-bromo-1-isobutyl-3-nitro-1H-pyrazole

  • MDL number MFCD31633770
  • Molecular Weight : 248.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart