PRIH8BA05A11 Princeton BioMolecular Research, Inc.

3-bromo-1-isobutyl-1H-pyrazol-5-amine

  • MDL number MFCD31633782
  • Molecular Weight : 218.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart