PRIH8BA05A15 Princeton BioMolecular Research, Inc.

1-ethyl-3-iodo-1H-pyrazol-5-amine

  • MDL number MFCD31633786
  • Molecular Weight : 237.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart