PRIH8BA05A29 Princeton BioMolecular Research, Inc.

2-(5-iodo-3-nitro-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD31633815
  • Molecular Weight : 311.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart