PRIH8BA05A45 Princeton BioMolecular Research, Inc.

6-methyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidin-2-amine

  • MDL number MFCD31633867
  • Molecular Weight : 216.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart